Gene Transcripts

Gene Symbol
GIMD1
Entrez Gene ID
100507096
Gene Name
GIMAP family P-loop NTPase domain containing 1
Gene Aliases
Chromosome Location
Chr. 4: 106357335 - 106368788 on Build GRCh38
UniGene ID
Hs.722750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195138.1 NP_001182067.1 2 411
XM_011531515.1 XP_011529817.1 3 492
GenBank mRNA DN831977.1 - 2 178

View Full Product Details