Gene Transcripts

Gene Symbol
NPAS4
Entrez Gene ID
266743
Gene Name
neuronal PAS domain protein 4
Gene Aliases
Le-PAS, NXF, PASD10, bHLHe79
Chromosome Location
Chr. 11: 66409158 - 66426707 on Build GRCh38
UniGene ID
Hs.256036
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017017537.1 XP_016873026.1 2 1228


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000526186.1 - 1 93 Chr. 11 - 66410672 - 66417127 [+]

View Full Product Details