Gene Transcripts
- Gene Symbol
- OVOL1
- Entrez Gene ID
- 5017
- Gene Name
- ovo like transcriptional repressor 1
- Gene Aliases
- HOVO1
- Chromosome Location
- Chr. 11: 65787034 - 65797219 on Build GRCh38
- UniGene ID
- Hs.134434
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AW207443.1 |
- |
1 |
122 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000531155.1 |
- |
1 |
105 |
|
Chr. 11 - 65795946 - 65797219 [-] |
View Full Product Details