Gene Transcripts

Gene Symbol
OVOL1
Entrez Gene ID
5017
Gene Name
ovo like transcriptional repressor 1
Gene Aliases
HOVO1
Chromosome Location
Chr. 11: 65787034 - 65797219 on Build GRCh38
UniGene ID
Hs.134434
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AW207443.1 - 1 145


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000531155.1 - 1 128 Chr. 11 - 65795946 - 65797219 [-]

View Full Product Details