Gene Transcripts

Gene Symbol
BLACAT1
Entrez Gene ID
101669762
Gene Name
bladder cancer associated transcript 1 (non-protein coding)
Gene Aliases
LINC00912, linc-UBC1, onco-lncRNA-30
Chromosome Location
Chr. 1: 205434886 - 205456086 on Build GRCh38
UniGene ID
Hs.666219
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_103783.1 2 2482
GenBank mRNA AK022594.1 - 1 2235
BX640973.1 - 1 2863

View Full Product Details