Gene Transcripts

Gene Symbol
Miat
Entrez Gene ID
330166
Gene Name
myocardial infarction associated transcript (non-protein coding)
Gene Aliases
3632434I06, A230057G18Rik, Rncr2, gomafu
Chromosome Location
Chr. 5: 112213228 - 112228948 on Build GRCm38
UniGene ID
Mm.44854
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_003718.2 7 3051
NR_033657.1 7 3095
GenBank mRNA AB300594.1 - 7 3038
AK053540.1 - 7 3039

View Full Product Details