Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_054066.4 | NP_473407.2 | 13, 14 | 2048 | |
GenBank mRNA | AF435950.1 | - | 13, 14 | 2053 | |
AK006672.1 | - | 13, 14 | 2093 | ||
BC106767.1 | - | 13, 14 | 2043 | 40039566 |