Gene Transcripts

Gene Symbol
Plxnb3
Entrez Gene ID
140571
Gene Name
plexin B3
Gene Aliases
AI451018, Plxn6
Chromosome Location
Chr. X: 73756559 - 73772510 on Build GRCm38
UniGene ID
Mm.275600
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019587.2 NP_062533.2 28 4875
GenBank mRNA AK122467.1 - 22 4103
AK138168.1 - 24 3880
AK138631.1 - 22 4056
AK140240.1 - 26 5057

View Full Product Details