Gene Transcripts

Gene Symbol
PAX7
Entrez Gene ID
5081
Gene Name
paired box 7
Gene Aliases
HUP1, PAX7B, RMS2
Chromosome Location
Chr. 1: 18631006 - 18748866 on Build GRCh38
UniGene ID
Hs.113253
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135254.1 NP_001128726.1 5 1322
NM_002584.2 NP_002575.1 5 1322
NM_013945.2 NP_039236.1 5 1316
GenBank mRNA BC121165.2 - 5 798
BC121166.2 - 5 798 40121584
DQ322591.1 - 5 807
X96743.1 - 5 1323
Z35141.1 - 5 826

View Full Product Details