Gene Transcripts

Gene Symbol
Tmprss7
Entrez Gene ID
208171
Gene Name
transmembrane serine protease 7
Gene Aliases
B230219I23Rik, Gm1748
Chromosome Location
Chr. 16: 45656309 - 45698553 on Build GRCm38
UniGene ID
Mm.186306
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172455.3 NP_766043.3 9 1122
GenBank mRNA AK045663.1 - 7 872
DQ061860.1 - 9 1123

View Full Product Details