Gene Transcripts

Gene Symbol
Tmem18
Entrez Gene ID
211986
Gene Name
transmembrane protein 18
Gene Aliases
BC024093
Chromosome Location
Chr. 12: 30584443 - 30591219 on Build GRCm38
UniGene ID
Mm.34035
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172049.2 NP_742046.2 4, 5 397
GenBank mRNA AK133493.1 - 4, 5 397
AK160817.1 - 4, 5 389
AK166323.1 - 4, 5 414
AK170496.1 - 4, 5 380
BC024093.1 - 4, 5 369 5116657

View Full Product Details