Gene Transcripts

Gene Symbol
Zswim4
Entrez Gene ID
212168
Gene Name
zinc finger SWIM-type containing 4
Gene Aliases
D330002I24, E130119J17Rik
Chromosome Location
Chr. 8: 84210942 - 84237042 on Build GRCm38
UniGene ID
Mm.117094
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172503.3 NP_766091.2 14 3265
GenBank mRNA AK052168.1 - 14 3279
AK053663.1 - 14 3279
AK131119.1 - 9 2123
BC016518.1 - 4 1082
BC058080.1 - 14 3265 6822376

View Full Product Details