Gene Transcripts

Gene Symbol
Ccdc92
Entrez Gene ID
215707
Gene Name
coiled-coil domain containing 92
Gene Aliases
AW050178, D5Bwg0834e
Chromosome Location
Chr. 5: 124834432 - 124862221 on Build GRCm38
UniGene ID
Mm.389693
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144819.2 NP_659068.1 2, 3 357
GenBank mRNA AK028192.1 - 2, 3 334
AK136203.1 - 2, 3 523
AK165371.1 - 2, 3 357
BC021492.1 - 2, 3 301 5345674

View Full Product Details