Gene Transcripts

Gene Symbol
Mgl2
Entrez Gene ID
216864
Gene Name
macrophage galactose N-acetyl-galactosamine specific lectin 2
Gene Aliases
CD301b
Chromosome Location
Chr. 11: 70130125 - 70137542 on Build GRCm38
UniGene ID
Mm.222465
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145137.2 NP_660119.1 10 1285
GenBank mRNA AK085751.1 - 8 1289
AY103461.1 - 10 1222
BC106833.1 - 9 1211 40044847
BC110475.2 - 9 1211 40045765
BC111520.1 - 9 1211 40042306
BY544307.1 - 1 189
EU010011.1 - 9 1364

View Full Product Details