Gene Transcripts

Gene Symbol
Rapgef3
Entrez Gene ID
223864
Gene Name
Rap guanine nucleotide exchange factor (GEF) 3
Gene Aliases
2310016P22Rik, 9330170P05Rik, Epac, Epac1
Chromosome Location
Chr. 15: 97744770 - 97768375 on Build GRCm38
UniGene ID
Mm.24028
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001177810.1 NP_001171281.1 9 1119
NM_001177811.1 NP_001171282.1 9 1119
NM_144850.2 NP_659099.2 9 1119
GenBank mRNA AK044566.1 - 9 1042
AK142594.1 - 9 1013
AK169430.1 - 9 1118
BC020532.1 - 9 1131 4236136

View Full Product Details