Gene Transcripts
- Gene Symbol
- D17Wsu92e
- Entrez Gene ID
- 224647
- Gene Name
- DNA segment, Chr 17, Wayne State University 92, expressed
- Gene Aliases
- AU020189, C030010A15, C80239
- Chromosome Location
- Chr. 17: 27751232 - 27820542 on Build GRCm38
- UniGene ID
- Mm.196577
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033279.3 |
NP_001028451.2
|
1, 2 |
374 |
|
NM_001044719.2 |
NP_001038184.1
|
1, 2 |
374 |
|
NM_001271511.1 |
NP_001258440.1
|
1, 2 |
374 |
|
GenBank mRNA |
AK143016.1 |
- |
1, 2 |
287 |
|
AK161603.1 |
- |
1, 2 |
375 |
|
BC132324.1 |
- |
2, 3 |
300 |
40130601 |
BC132326.1 |
- |
2, 3 |
300 |
40130603 |
BC137573.1 |
- |
1, 2 |
374 |
8860592 |
BC137576.1 |
- |
1, 2 |
374 |
8860595 |
BC145403.1 |
- |
1, 2 |
374 |
9053961 |
BY755073.1 |
- |
1, 2 |
300 |
|
CX220817.1 |
- |
1, 2 |
149 |
|
View Full Product Details