Gene Transcripts

Gene Symbol
Abcf1
Entrez Gene ID
224742
Gene Name
ATP-binding cassette, sub-family F (GCN20), member 1
Gene Aliases
AU041969, Abc50, D17Wsu166e, GCN20
Chromosome Location
Chr. 17: 35956816 - 35969800 on Build GRCm38
UniGene ID
Mm.329022
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013854.1 NP_038882.1 10 821
GenBank mRNA BC046965.1 - 10 792
BC063094.1 - 10 821 6516514

View Full Product Details