Gene Transcripts

Gene Symbol
Catsper1
Entrez Gene ID
225865
Gene Name
cation channel, sperm associated 1
Gene Aliases
Catsper, KSper
Chromosome Location
Chr. 19: 5335602 - 5344281 on Build GRCm38
UniGene ID
Mm.87321
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139301.2 NP_647462.1 1 385
GenBank mRNA AF407332.1 - 1 385

View Full Product Details