Gene Transcripts

Gene Symbol
Trpm6
Entrez Gene ID
225997
Gene Name
transient receptor potential cation channel, subfamily M, member 6
Gene Aliases
CHAK2, TRPM6/CHAK2
Chromosome Location
Chr. 19: 18730178 - 18892516 on Build GRCm38
UniGene ID
Mm.215171
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153417.1 NP_700466.1 14 1586
GenBank mRNA AK080899.1 - 14 1585
AY135644.1 - 14 1586
BC145223.1 - 14 1578

View Full Product Details