Gene Transcripts

Gene Symbol
Sbspon
Entrez Gene ID
226866
Gene Name
somatomedin B and thrombospondin, type 1 domain containing
Gene Aliases
Gm106, Rpesp
Chromosome Location
Chr. 1: 15853862 - 15892722 on Build GRCm38
UniGene ID
Mm.131665
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033288.3 NP_001028460.1 4 717
GenBank mRNA AK137144.1 - 4 718
AK143251.1 - 4 691
BC147194.1 - 4 704 9056240
BC147195.1 - 4 704 9056224

View Full Product Details