Gene Transcripts

Gene Symbol
Ralgapb
Entrez Gene ID
228850
Gene Name
Ral GTPase activating protein, beta subunit (non-catalytic)
Gene Aliases
9330195P16, AI507211, B230339M05Rik, mKIAA1219, p170
Chromosome Location
Chr. 2: 158409829 - 158499253 on Build GRCm38
UniGene ID
Mm.398185
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291137.1 NP_001278066.1 11 1761
NM_001291138.1 NP_001278067.1 10 1725
GenBank mRNA AK122470.1 - 5 544
BC138377.1 - 11 1663 8861397
BC145260.1 - 10 1627
CK638301.1 - 4 443

View Full Product Details