Gene Transcripts

Gene Symbol
Tapt1
Entrez Gene ID
231225
Gene Name
transmembrane anterior posterior transformation 1
Gene Aliases
4932414K18Rik
Chromosome Location
Chr. 5: 44175156 - 44229281 on Build GRCm38
UniGene ID
Mm.30688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173764.3 NP_776125.2 11, 12 1225
GenBank mRNA BC096038.1 - 8, 9 704
BC110670.1 - 10, 11 946

View Full Product Details