Gene Transcripts

Gene Symbol
POLD2
Entrez Gene ID
5425
Gene Name
DAN polymerase delta 2, accessory subunit
Gene Aliases
Chromosome Location
Chr. 7: 44114680 - 44123668 on Build GRCh38
UniGene ID
Hs.306791
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127218.2 NP_001120690.1 9 1337
NM_001256879.1 NP_001243808.1 10 1698
NM_006230.3 NP_006221.2 9 1164
XM_011515431.1 XP_011513733.1 9 1160
GenBank mRNA AK292347.1 - 10 1698
AK298168.1 - 9 1028
AK312294.1 - 9 1297
BC000459.2 - 9 1127 2822169
BC030001.2 - 8 1104
U21090.1 - 9 1126

View Full Product Details