Gene Transcripts

Gene Symbol
Fhod1
Entrez Gene ID
234686
Gene Name
formin homology 2 domain containing 1
Gene Aliases
BC017144, FHOS, FHOS1
Chromosome Location
Chr. 8: 105329160 - 105347970 on Build GRCm38
UniGene ID
Mm.87724
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177699.4 NP_808367.2 8 936
GenBank mRNA AB041045.1 - 8 830
AK030737.1 - 8 898
BC060654.1 - 8 919 30354865

View Full Product Details