Gene Transcripts

Gene Symbol
Mmp27
Entrez Gene ID
234911
Gene Name
matrix metallopeptidase 27
Gene Aliases
Gm180
Chromosome Location
Chr. 9: 7571396 - 7581885 on Build GRCm38
UniGene ID
Mm.334940
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030289.1 NP_001025460.1 9 1588
GenBank mRNA AK155813.1 - 10 1575
AK159182.1 - 9 1343

View Full Product Details