Gene Transcripts

Gene Symbol
Prss42
Entrez Gene ID
235628
Gene Name
protease, serine 42
Gene Aliases
Tessp2
Chromosome Location
Chr. 9: 110797261 - 110803744 on Build GRCm38
UniGene ID
Mm.292285
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153099.1 NP_694739.1 3 494
GenBank mRNA AB052292.1 - 3 494
AK133402.1 - 3 359
BC060984.1 - 3 513 6774705

View Full Product Details