Gene Transcripts

Gene Symbol
Ccdc55
Entrez Gene ID
237859
Gene Name
coiled-coil domain containing 55
Gene Aliases
8030451K07, AI851076, AL022898, NSpr70, NSrp70, Nsrp1
Chromosome Location
Chr. 11: 77044292 - 77078437 on Build GRCm38
UniGene ID
Mm.156195
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012309.2 NP_001012309.1 7 1394
GenBank mRNA AK078762.1 - 7 1380
AK168573.1 - 7 1380
BC089560.1 - 7 1427 30300075

View Full Product Details