Gene Transcripts

Gene Symbol
Tubal3
Entrez Gene ID
238463
Gene Name
tubulin, alpha-like 3
Gene Aliases
Chromosome Location
Chr. 13: 3924635 - 3935277 on Build GRCm38
UniGene ID
Mm.313659
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033879.3 NP_001029051.2 4 469
GenBank mRNA AK135971.1 - 4 470
BC147507.1 - 4 462 9056442

View Full Product Details