Gene Transcripts

Gene Symbol
Zpld1
Entrez Gene ID
239852
Gene Name
zona pellucida like domain containing 1
Gene Aliases
9430016A21Rik
Chromosome Location
Chr. 16: 55224125 - 55582475 on Build GRCm38
UniGene ID
Mm.245045
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178720.4 NP_848835.1 3 216
GenBank mRNA AK034628.1 - 3 217
AK047981.1 - 2 126
AK052599.1 - 4 326
BC116254.1 - 3 185 40093631
BC116255.1 - 3 185 40093632

View Full Product Details