Gene Transcripts

Gene Symbol
PPP3CA
Entrez Gene ID
5530
Gene Name
protein phosphatase 3 catalytic subunit alpha
Gene Aliases
CALN, CALNA, CALNA1, CCN1, CNA1, PPP2B
Chromosome Location
Chr. 4: 101023430 - 101347471 on Build GRCh38
UniGene ID
Hs.435512
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000944.4 NP_000935.1 5 1304
NM_001130691.1 NP_001124163.1 5 1304
NM_001130692.1 NP_001124164.1 5 1304
XM_017008365.1 XP_016863854.1 5 608
GenBank mRNA AB451338.1 - 5 629
AB451487.1 - 5 629
AK290532.1 - 5 1166
AL353950.1 - 5 1035
AY904364.1 - 4 474
BC025714.1 - 5 738 5227691
EU192652.1 - 5 1304
KU178269.1 - 5 629
KU178270.1 - 5 629
L14778.1 - 5 776

View Full Product Details