Gene Transcripts

Gene Symbol
PRG2
Entrez Gene ID
5553
Gene Name
proteoglycan 2, pro eosinophil major basic protein
Gene Aliases
BMPG, MBP, MBP1, proMBP
Chromosome Location
Chr. 11: 57386787 - 57390657 on Build GRCh38
UniGene ID
Hs.512633
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243245.2 NP_001230174.1 2, 3 142
NM_001302926.1 NP_001289855.1 2, 3 138
NM_001302927.1 NP_001289856.1 2, 3 255
NM_002728.5 NP_002719.3 2, 3 142
GenBank mRNA AK312195.1 - 2, 3 131
AU135652.1 - 2, 3 244
BC005929.1 - 2, 3 135 4043815
CB997424.1 - 2, 3 162
CR450311.1 - 1, 2 68
DA841825.1 - 2, 3 132
DQ846874.1 - 1, 2 68
M35670.1 - 2, 3 116
M36805.1 - 2, 3 136
X14088.1 - 2, 3 138
X65787.1 - 2, 3 142
Y00809.1 - 2, 3 136
Z26248.1 - 9, 10 925

View Full Product Details