Gene Transcripts

Gene Symbol
Eda2r
Entrez Gene ID
245527
Gene Name
ectodysplasin A2 receptor
Gene Aliases
9430060M22Rik, TNFRSF27, Xedar
Chromosome Location
Chr. X: 97333837 - 97377218 on Build GRCm38
UniGene ID
Mm.189270
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161432.1 NP_001154904.1 6, 7 785
NM_001161433.1 NP_001154905.1 6, 7 740
NM_175540.5 NP_780749.3 6, 7 740
GenBank mRNA AK034909.1 - 6, 7 704
AK049134.1 - 6, 7 747
BC106845.1 - 6, 7 726 40045945

View Full Product Details