Gene Transcripts

Gene Symbol
Tex10
Entrez Gene ID
269536
Gene Name
testis expressed gene 10
Gene Aliases
2610206N19Rik, 2810462N03Rik, BC006867
Chromosome Location
Chr. 4: 48430858 - 48473464 on Build GRCm38
UniGene ID
Mm.272025
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172304.3 NP_758508.2 7 1774
GenBank mRNA AK132870.1 - 7 1775
AK141288.1 - 6 1553
BC006867.1 - 4 1021
BC151176.1 - 6 1525 9088065
BC151177.1 - 6 1525 9088066

View Full Product Details