Gene Transcripts

Gene Symbol
Cntn4
Entrez Gene ID
269784
Gene Name
contactin 4
Gene Aliases
9630050B05, Axcam, BIG-2
Chromosome Location
Chr. 6: 105677651 - 106700141 on Build GRCm38
UniGene ID
Mm.321683
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109749.1 NP_001103219.1 13 1608
NM_001109751.1 NP_001103221.1 12 1696
NM_173004.3 NP_766592.2 13 1608
GenBank mRNA AK036262.1 - 13 1578
AK043507.1 - 10 1553
AK044694.1 - 3 356
AK163520.1 - 10 1401
AK173340.1 - 6 727
BC046975.1 - 10 1230
BC115766.1 - 13 1498 40060288

View Full Product Details