Gene Transcripts

Gene Symbol
Elmod1
Entrez Gene ID
270162
Gene Name
ELMO/CED-12 domain containing 1
Gene Aliases
4831417L10, AI428506, AI834991
Chromosome Location
Chr. 9: 53911457 - 53975310 on Build GRCm38
UniGene ID
Mm.259791
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177769.4 NP_808437.2 11 1189
GenBank mRNA AK132239.1 - 11 1190
AK164094.1 - 341
BC049160.1 - 9 849

View Full Product Details