Gene Transcripts

Gene Symbol
Pcdh19
Entrez Gene ID
279653
Gene Name
protocadherin 19
Gene Aliases
B530002L05Rik, Gm717, mKIAA1313
Chromosome Location
Chr. X: 133582860 - 133688993 on Build GRCm38
UniGene ID
Mm.39738
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105245.1 NP_001098715.1 6 5118
NM_001105246.1 NP_001098716.1 5 4974
GenBank mRNA AK122492.1 - 6 2861
BC118529.1 - 6 3163 40098492

View Full Product Details