Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252094.1 | NP_001239023.1 | 3, 4 | 728 | |
NM_001252095.1 | NP_001239024.1 | 3, 4 | 709 | ||
NM_001252096.1 | NP_001239025.1 | 2, 3 | 618 | ||
NM_177101.5 | NP_796075.2 | 2, 3 | 599 | ||
GenBank mRNA | AK132096.1 | - | 3, 4 | 596 | |
BC048711.1 | - | 2, 3 | 491 | 6328428 | |
BC065807.1 | - | 2, 3 | 515 | 3373106 | |
DN174886.1 | - | 2, 3 | 492 |