Gene Transcripts

Gene Symbol
Tnpo3
Entrez Gene ID
320938
Gene Name
transportin 3
Gene Aliases
5730544L10Rik, C430013M08Rik, C81142, D6Ertd313e, Trn-SR, mKIAA4133
Chromosome Location
Chr. 6: 29540827 - 29610110 on Build GRCm38
UniGene ID
Mm.276381
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177296.4 NP_796270.2 20 2576
GenBank mRNA AK077714.1 - 4 424
AK220184.1 - 13 1479
BC023273.1 - 8 761
BC052756.1 - 9 897
BC064646.1 - 20 2576 6313452
BC075678.1 - 20 2789 30619729

View Full Product Details