Gene Transcripts

Gene Symbol
Lrrn4
Entrez Gene ID
320974
Gene Name
leucine rich repeat neuronal 4
Gene Aliases
B430119L13Rik, Nlrr4
Chromosome Location
Chr. 2: 132866194 - 132894009 on Build GRCm38
UniGene ID
Mm.357342
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177303.4 NP_796277.2 5 1904
GenBank mRNA AK080889.1 - 5 1905
AK085246.1 - 5 1934
AY325108.1 - 5 1893

View Full Product Details