Gene Transcripts

Gene Symbol
Ubtd2
Entrez Gene ID
327900
Gene Name
ubiquitin domain containing 2
Gene Aliases
4930571L24, 9630054F20Rik, AI645720
Chromosome Location
Chr. 11: 32455372 - 32518709 on Build GRCm38
UniGene ID
Mm.260956
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173784.3 NP_776145.2 3 548
GenBank mRNA AK076953.1 - 3 550
BC043664.1 - 1 43
BC057026.1 - 3 548 6825513

View Full Product Details