Gene Transcripts

Gene Symbol
Miat
Entrez Gene ID
330166
Gene Name
myocardial infarction associated transcript (non-protein coding)
Gene Aliases
3632434I06, A230057G18Rik, Rncr2, gomafu
Chromosome Location
Chr. 5: 112213228 - 112228948 on Build GRCm38
UniGene ID
Mm.44854
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB294520.1 - 2 126
AB294521.1 - 2 126
AB294522.1 - 2 126
AB294525.1 - 2 126
AB294526.1 - 2 126
AB294527.1 - 2 126
AB300594.1 - 2 363
AK038726.1 - 2 377
AK053540.1 - 2 363

View Full Product Details