Gene Transcripts
- Gene Symbol
- Miat
- Entrez Gene ID
- 330166
- Gene Name
- myocardial infarction associated transcript (non-protein coding)
- Gene Aliases
- 3632434I06, A230057G18Rik, Rncr2, gomafu
- Chromosome Location
- Chr. 5: 112213228 - 112228948 on Build GRCm38
- UniGene ID
- Mm.44854
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB294520.1 |
- |
4, 5 |
384 |
|
AB294521.1 |
- |
3, 4 |
281 |
|
AB294522.1 |
- |
3, 4 |
273 |
|
AB294523.1 |
- |
2, 3 |
261 |
|
AB294524.1 |
- |
2, 3 |
225 |
|
AB294525.1 |
- |
3, 4 |
317 |
|
AB294526.1 |
- |
3, 4 |
281 |
|
AB294527.1 |
- |
3, 4 |
273 |
|
AB294528.1 |
- |
2, 3 |
225 |
|
AB300594.1 |
- |
3, 4 |
510 |
|
AK038726.1 |
- |
3, 4 |
568 |
|
AK053540.1 |
- |
3, 4 |
511 |
|
View Full Product Details