Gene Transcripts

Gene Symbol
Thsd7a
Entrez Gene ID
330267
Gene Name
thrombospondin, type I, domain containing 7A
Gene Aliases
Gm837
Chromosome Location
Chr. 6: 12311608 - 12749253 on Build GRCm38
UniGene ID
Mm.11989
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164805.1 NP_001158277.1 28 5198
GenBank mRNA AK133324.1 - 23 3278
AK164834.1 - 2 89
AK173072.1 - 28 4969

View Full Product Details