Gene Transcripts

Gene Symbol
Frem3
Entrez Gene ID
333315
Gene Name
Fras1 related extracellular matrix protein 3
Gene Aliases
Gm795, NV2
Chromosome Location
Chr. 8: 80611039 - 80695557 on Build GRCm38
UniGene ID
Mm.134296
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167898.1 NP_001161370.1 1 3598

View Full Product Details