Gene Transcripts

Gene Symbol
Tas2r117
Entrez Gene ID
353166
Gene Name
taste receptor, type 2, member 117
Gene Aliases
T2R17, Tas2r17, mGR17, mt2r54
Chromosome Location
Chr. 6: 132802901 - 132803893 on Build GRCm38
UniGene ID
Mm.377890
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207021.1 NP_996904.1 1 268
GenBank mRNA BC132206.1 - 1 351
BC132208.1 - 1 351 40130485

View Full Product Details