Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006265.2 | NP_006256.1 | 8 | 1214 | |
GenBank mRNA | AK098521.1 | - | 4 | 458 | |
AK289505.1 | - | 8 | 991 | ||
BC050381.1 | - | 8 | 1054 | 6044010 | |
D38551.1 | - | 8 | 1110 | ||
X98294.1 | - | 7 | 926 |