Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142804.1 | NP_001136276.1 | 1 | 338 | |
NM_198636.3 | NP_941038.2 | 1 | 338 | ||
GenBank mRNA | AK033376.1 | - | 1 | 343 | |
AK163897.1 | - | 1 | 325 | ||
BC113198.1 | - | 1 | 302 | 40062399 | |
BY720548.1 | - | 1 | 338 |