Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026958.3 | NP_081234.2 | 3, 4 | 309 | |
GenBank mRNA | AJ293625.1 | - | 3 | 681 | |
AK007699.1 | - | 3, 4 | 296 | ||
AK028746.1 | - | 287 | |||
BC065396.1 | - | 3 | 681 | ||
BC100567.1 | - | 3, 4 | 274 | ||
BC116739.1 | - | 3, 4 | 296 | 40126058 | |
BC116741.1 | - | 3, 4 | 296 | 40126060 | |
BU850988.1 | - | 3, 4 | 312 | ||
CK372908.1 | - | 3, 4 | 279 |