Gene Transcripts

Gene Symbol
Nsl1
Entrez Gene ID
381318
Gene Name
NSL1, MIND kinetochore complex component, homolog (S. cerevisiae)
Gene Aliases
4833432M17Rik, AI451597, Gm105, Gm984
Chromosome Location
Chr. 1: 191063012 - 191084558 on Build GRCm38
UniGene ID
Mm.32504
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198654.3 NP_941056.3 2 310
GenBank mRNA AK029421.1 - 2 308
BC029086.1 - 2 300 3156934

View Full Product Details