Gene Transcripts

Gene Symbol
Kpna7
Entrez Gene ID
381686
Gene Name
karyopherin alpha 7 (importin alpha 8)
Gene Aliases
AW146299, Gm1055
Chromosome Location
Chr. 5: 144983727 - 145009636 on Build GRCm38
UniGene ID
Mm.332837
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013774.2 NP_001013796.2 8 1091
GenBank mRNA AK136027.1 - 4 609
AY950703.1 - 8 1092
FJ717332.1 - 6 858
FJ717333.1 - 7 921

View Full Product Details